Assay Details
| Gene Symbol: Lcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504283.2 | XP_003504331.1 | 5 - 6 | 955 | 73 | |
| XM_007644850.1 | XP_007643040.1 | 4 - 5 | 1446 | 73 | ||
| GenBank mRNA | JI875247.1 | - | 1 - 2 | 293 | 73 | |
More Information
Set Membership: |
|

Lcat
Genomic Map