Assay Details
| Gene Symbol: Mkl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507467.2 | XP_003507515.2 | 6 - 7 | 792 | 55 | |
| XM_007648062.1 | XP_007646252.1 | 8 - 9 | 935 | 55 | ||
| XM_007648063.1 | XP_007646253.1 | 7 - 8 | 713 | 55 | ||
| GenBank mRNA | JI869299.1 | - | 6 - 7 | 554 | 55 | |
| JP046634.1 | - | 6 - 7 | 554 | 55 | ||
More Information
Set Membership: |
|

Mkl1
Genomic Map