Assay Details
| Gene Symbol: Sh2d1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508143.1 | XP_003508191.1 | 1 - 2 | 130 | 86 | |
| XM_007648763.1 | XP_007646953.1 | 1 - 2 | 130 | 86 | ||
More Information
Set Membership: |
|

Sh2d1a
Genomic Map