Assay Details
| Gene Symbol: Tbc1d20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511491.2 | XP_003511539.1 | 6 - 7 | 886 | 58 | |
| GenBank mRNA | JI869980.1 | - | 6 - 7 | 731 | 58 | |
| JP038539.1 | - | 6 - 7 | 731 | 58 | ||
More Information
Set Membership: |
|

Tbc1d20
Genomic Map