Assay Details
| Gene Symbol: Cstb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515067.2 | XP_003515115.1 | 1 - 2 | 198 | 81 | |
| GenBank mRNA | JI866087.1 | - | 1 - 2 | 192 | 81 | |
| JP060891.1 | - | 1 - 2 | 192 | 81 | ||
More Information
Set Membership: |
|

Cstb
Genomic Map