Assay Details
| Gene Symbol: LOC100763859 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003516058.2 | XP_003516106.2 | 1 - 2 | 223 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100763859| Gene Symbol: LOC100763859 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003516058.2 | XP_003516106.2 | 1 - 2 | 223 | 74 | |
Set Membership: |
|