Assay Details
| Gene Symbol: Nr4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654639.1 | XP_007652829.1 | 6 - 7 | 2487 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nr4a1| Gene Symbol: Nr4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654639.1 | XP_007652829.1 | 6 - 7 | 2487 | 65 | |
Set Membership: |
|