Assay Details
| Gene Symbol: Fbxo31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007654068.1 | XP_007652258.1 | 6 - 7 | 1057 | 63 | |
| XM_007654115.1 | XP_007652305.1 | 9 - 10 | 1253 | 63 | ||
| XM_007654160.1 | XP_007652350.1 | 7 - 8 | 1093 | 63 | ||
More Information
Set Membership: |
|

Fbxo31
Genomic Map