Assay Details
| Gene Symbol: Slc25a41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642437.1 | XP_007640627.1 | 2 - 3 | 432 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a41| Gene Symbol: Slc25a41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642437.1 | XP_007640627.1 | 2 - 3 | 432 | 69 | |
Set Membership: |
|