Assay Details
| Gene Symbol: Cabyr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647159.1 | XP_007645349.1 | 3 - 4 | 1721 | 61 | |
| XM_007647160.1 | XP_007645350.1 | 4 - 5 | 3231 | 61 | ||
| XM_007647162.1 | XP_007645352.1 | 3 - 4 | 1718 | 61 | ||
More Information
Set Membership: |
|

Cabyr
Genomic Map