Assay Details
| Gene Symbol: Prickle3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505082.2 | XP_003505130.1 | 2 - 3 | 126 | 61 | |
| XM_007645608.1 | XP_007643798.1 | 2 - 3 | 126 | 61 | ||
| XM_007645609.1 | XP_007643799.1 | 2 - 3 | 126 | 61 | ||
More Information
Set Membership: |
|

Prickle3
Genomic Map