Assay Details
| Gene Symbol: Racgap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507843.2 | XP_003507891.1 | 15 - 16 | 1827 | 63 | |
| GenBank mRNA | JI865814.1 | - | 5 - 6 | 665 | 63 | |
| JP046518.1 | - | 5 - 6 | 665 | 63 | ||
More Information
Set Membership: |
|

Racgap1
Genomic Map