Assay Details
Gene Symbol: LOC100772476 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509461.2 | XP_003509509.1 | 1 - 2 | 240 | 90 | |
GenBank mRNA | JI878921.1 | - | 1 - 2 | 158 | 90 | |
JP039710.1 | - | 1 - 2 | 169 | 90 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
