Assay Details
| Gene Symbol: LOC100772476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509461.2 | XP_003509509.1 | 4 - 5 | 792 | 96 | |
| XM_007650054.1 | XP_007648244.1 | 3 - 4 | 1122 | 96 | ||
| GenBank mRNA | JI878921.1 | - | 4 - 5 | 710 | 96 | |
| JP039710.1 | - | 4 - 5 | 721 | 96 | ||
More Information
Set Membership: |
|

LOC100772476
Genomic Map