Assay Details
| Gene Symbol: LOC100772476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509461.2 | XP_003509509.1 | 7 - 8 | 1334 | 64 | |
| XM_007650054.1 | XP_007648244.1 | 6 - 7 | 1664 | 64 | ||
| GenBank mRNA | JI878921.1 | - | 7 - 8 | 1252 | 64 | |
| JP039710.1 | - | 7 - 8 | 1263 | 64 | ||
More Information
Set Membership: |
|

LOC100772476
Genomic Map