Assay Details
| Gene Symbol: LOC100750898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509835.1 | XP_003509883.1 | 1 - 2 | 35 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100750898| Gene Symbol: LOC100750898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509835.1 | XP_003509883.1 | 1 - 2 | 35 | 70 | |
Set Membership: |
|