Assay Details
| Gene Symbol: Prdx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652058.1 | XP_007650248.1 | 3 - 4 | 570 | 66 | |
| GenBank mRNA | JP056430.1 | - | 3 - 4 | 570 | 66 | |
More Information
Set Membership: |
|

Prdx4
Genomic Map