Assay Details
| Gene Symbol: Cmss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645369.1 | XP_007643559.1 | 5 - 6 | 405 | 68 | |
| GenBank mRNA | JP055802.1 | - | 3 - 4 | 288 | 68 | |
More Information
Set Membership: |
|

Cmss1
Genomic Map