Assay Details
| Gene Symbol: Immp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497564.2 | XP_003497612.1 | 7 - 8 | 801 | 69 | |
| XM_007613937.1 | XP_007612127.1 | 7 - 8 | 839 | 69 | ||
| XM_007613952.1 | XP_007612142.1 | 8 - 9 | 1095 | 69 | ||
| XM_007613960.1 | XP_007612150.1 | 8 - 9 | 1057 | 69 | ||
More Information
Set Membership: |
|

Immp1l
Genomic Map