Assay Details
| Gene Symbol: Taf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498375.2 | XP_003498423.1 | 4 - 5 | 559 | 66 | |
| XM_007626813.1 | XP_007625003.1 | 4 - 5 | 703 | 66 | ||
| XM_007626820.1 | XP_007625010.1 | 5 - 6 | 675 | 66 | ||
| XM_007626827.1 | XP_007625017.1 | 5 - 6 | 817 | 66 | ||
| GenBank mRNA | JI864725.1 | - | 3 - 4 | 446 | 66 | |
| JP058536.1 | - | 3 - 4 | 446 | 66 | ||
More Information
Set Membership: |
|

Taf12
Genomic Map