Assay Details
| Gene Symbol: Hscb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501710.2 | XP_003501758.1 | 2 - 3 | 374 | 64 | |
| GenBank mRNA | JI867316.1 | - | 2 - 3 | 371 | 64 | |
| JP054176.1 | - | 2 - 3 | 371 | 64 | ||
More Information
Set Membership: |
|

Hscb
Genomic Map