Assay Details
| Gene Symbol: Rem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502332.1 | XP_003502380.1 | 1 - 2 | 343 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rem1| Gene Symbol: Rem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502332.1 | XP_003502380.1 | 1 - 2 | 343 | 60 | |
Set Membership: |
|