Assay Details
| Gene Symbol: Cdh9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505285.2 | XP_003505333.1 | 8 - 9 | 1895 | 73 | |
| XM_007645810.1 | XP_007644000.1 | 7 - 8 | 1776 | 73 | ||
| GenBank mRNA | JI865970.1 | - | 6 - 7 | 989 | 73 | |
| JP051460.1 | - | 6 - 7 | 989 | 73 | ||
More Information
Set Membership: |
|

Cdh9
Genomic Map