Assay Details
| Gene Symbol: LOC100773817 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507765.2 | XP_003507813.1 | 6 - 7 | 566 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100773817| Gene Symbol: LOC100773817 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507765.2 | XP_003507813.1 | 6 - 7 | 566 | 64 | |
Set Membership: |
|