Assay Details
| Gene Symbol: Park7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003513304.2 | XP_003513352.1 | 2 - 3 | 318 | 60 | |
| GenBank mRNA | JI864097.1 | - | 2 - 3 | 327 | 60 | |
| JP051795.1 | - | 2 - 3 | 327 | 60 | ||
More Information
Set Membership: |
|

Park7
Genomic Map