Assay Details
| Gene Symbol: Phkg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652273.1 | XP_007650463.1 | 7 - 8 | 1033 | 59 | |
| GenBank mRNA | JI863283.1 | - | 4 - 5 | 468 | 59 | |
| JP062048.1 | - | 4 - 5 | 468 | 59 | ||
More Information
Set Membership: |
|

Phkg2
Genomic Map