Assay Details
| Gene Symbol: LOC100773571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496915.2 | XP_003496963.1 | 3 - 4 | 435 | 64 | |
| XM_007654318.1 | XP_007652508.1 | 3 - 4 | 271 | 64 | ||
| GenBank mRNA | JI877839.1 | - | 3 - 4 | 341 | 64 | |
More Information
Set Membership: |
|

LOC100773571
Genomic Map