Assay Details
| Gene Symbol: LOC100762694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514393.2 | XP_003514441.1 | 6 - 7 | 992 | 81 | |
| XM_007654545.1 | XP_007652735.1 | 6 - 7 | 879 | 81 | ||
| XM_007654547.1 | XP_007652737.1 | 6 - 7 | 836 | 81 | ||
| GenBank mRNA | JI893904.1 | - | 6 - 7 | 971 | 81 | |
| JP046820.1 | - | 6 - 7 | 971 | 81 | ||
More Information
Set Membership: |
|

LOC100762694
Genomic Map