Assay Details
| Gene Symbol: LOC100772712 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515078.1 | XP_003515126.1 | 5 - 6 | 2770 | 66 | |
| XM_003515079.1 | XP_003515127.1 | 3 - 4 | 1426 | 66 | ||
| XM_003515080.1 | XP_003515128.1 | 4 - 5 | 2755 | 66 | ||
| XM_003515081.1 | XP_003515129.1 | 2 - 3 | 394 | 66 | ||
More Information
Set Membership: |
|

LOC100772712
Genomic Map