Assay Details
| Gene Symbol: Crtc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499105.2 | XP_003499153.1 | 7 - 8 | 555 | 57 | |
| GenBank mRNA | JI870659.1 | - | 3 - 4 | 222 | 57 | |
| JP044761.1 | - | 3 - 4 | 219 | 57 | ||
More Information
Set Membership: |
|

Crtc1
Genomic Map