Assay Details
| Gene Symbol: LOC100753660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510276.2 | XP_003510324.1 | 2 - 3 | 170 | 82 | |
| XM_007650923.1 | XP_007649113.1 | 3 - 4 | 298 | 82 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100753660| Gene Symbol: LOC100753660 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510276.2 | XP_003510324.1 | 2 - 3 | 170 | 82 | |
| XM_007650923.1 | XP_007649113.1 | 3 - 4 | 298 | 82 | ||