Assay Details
| Gene Symbol: Parp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647711.1 | XP_007645901.1 | 2 - 3 | 438 | 54 | |
| XM_007647712.1 | XP_007645902.1 | 2 - 3 | 438 | 54 | ||
More Information
Set Membership: |
|

Parp16
Genomic Map