Assay Details
| Gene Symbol: Deaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650430.1 | XP_007648620.1 | 5 - 6 | 966 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Deaf1| Gene Symbol: Deaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650430.1 | XP_007648620.1 | 5 - 6 | 966 | 60 | |
Set Membership: |
|