Assay Details
| Gene Symbol: Paip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645443.1 | XP_007643633.1 | 3 - 4 | 430 | 68 | |
| XM_007645444.1 | XP_007643634.1 | 3 - 4 | 415 | 68 | ||
| GenBank mRNA | JI863066.1 | - | 2 - 3 | 376 | 68 | |
More Information
Set Membership: |
|

Paip1
Genomic Map