Assay Details
| Gene Symbol: Csrp2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496467.2 | XP_003496515.2 | 6 - 7 | 1694 | 75 | |
| XM_007650434.1 | XP_007648624.1 | 6 - 7 | 1421 | 75 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csrp2bp| Gene Symbol: Csrp2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496467.2 | XP_003496515.2 | 6 - 7 | 1694 | 75 | |
| XM_007650434.1 | XP_007648624.1 | 6 - 7 | 1421 | 75 | ||