Assay Details
| Gene Symbol: Cib2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497896.2 | XP_003497944.1 | 5 - 6 | 677 | 75 | |
| XM_003497897.2 | XP_003497945.1 | 5 - 6 | 780 | 75 | ||
| XM_007619246.1 | XP_007617436.1 | 4 - 5 | 558 | 75 | ||
| GenBank mRNA | JI879528.1 | - | 4 - 5 | 575 | 75 | |
| JP043662.1 | - | 4 - 5 | 497 | 75 | ||
More Information
Set Membership: |
|

Cib2
Genomic Map