Assay Details
| Gene Symbol: Rbbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501200.2 | XP_003501248.1 | 7 - 8 | 896 | 76 | |
| XM_007641896.1 | XP_007640086.1 | 6 - 7 | 868 | 76 | ||
| XM_007641897.1 | XP_007640087.1 | 7 - 8 | 903 | 76 | ||
| GenBank mRNA | JI863883.1 | - | 7 - 8 | 863 | 76 | |
| JP058802.1 | - | 7 - 8 | 863 | 76 | ||
More Information
Set Membership: |
|

Rbbp5
Genomic Map