Assay Details
| Gene Symbol: Pmvk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501600.2 | XP_003501648.1 | 2 - 3 | 384 | 75 | |
| GenBank mRNA | JI894467.1 | - | 2 - 3 | 225 | 75 | |
| JP040453.1 | - | 2 - 3 | 225 | 75 | ||
More Information
Set Membership: |
|

Pmvk
Genomic Map