Assay Details
| Gene Symbol: Tab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507917.2 | XP_003507965.1 | 8 - 9 | 2267 | 65 | |
| XM_007648507.1 | XP_007646697.1 | 10 - 11 | 2448 | 65 | ||
| XM_007648508.1 | XP_007646698.1 | 9 - 10 | 2354 | 65 | ||
| GenBank mRNA | JI869027.1 | - | 5 - 6 | 1897 | 65 | |
| JP059362.1 | - | 5 - 6 | 1897 | 65 | ||
More Information
Set Membership: |
|

Tab3
Genomic Map