Assay Details
| Gene Symbol: Rpl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509460.2 | XP_003509508.1 | 4 - 5 | 580 | 60 | |
| GenBank mRNA | JI879834.1 | - | 4 - 5 | 560 | 60 | |
| JP046153.1 | - | 4 - 5 | 559 | 60 | ||
More Information
Set Membership: |
|

Rpl3
Genomic Map