Assay Details
| Gene Symbol: Epgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512369.2 | XP_003512417.1 | 1 - 2 | 35 | 68 | |
| XM_007652863.1 | XP_007651053.1 | 1 - 2 | 35 | 68 | ||
| XM_007652864.1 | XP_007651054.1 | 1 - 2 | 35 | 68 | ||
| XM_007652865.1 | XP_007651055.1 | 1 - 2 | 35 | 68 | ||
| XM_007652866.1 | XP_007651056.1 | 1 - 2 | 35 | 68 | ||
More Information
Set Membership: |
|

Epgn
Genomic Map