Assay Details
| Gene Symbol: Scn4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515273.1 | XP_003515321.1 | 3 - 4 | 464 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scn4b| Gene Symbol: Scn4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515273.1 | XP_003515321.1 | 3 - 4 | 464 | 68 | |
Set Membership: |
|