Assay Details
| Gene Symbol: Cd2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003515697.2 | XP_003515745.1 | 2 - 3 | 193 | 74 | |
| XM_007607822.1 | XP_007606012.1 | 2 - 3 | 200 | 74 | ||
| XM_007607823.1 | XP_007606013.1 | 2 - 3 | 197 | 74 | ||
More Information
Set Membership: |
|

Cd2bp2
Genomic Map