Assay Details
| Gene Symbol: Tmem25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007651141.1 | XP_007649331.1 | 5 - 6 | 1073 | 61 | |
| XM_007651142.1 | XP_007649332.1 | 4 - 5 | 941 | 61 | ||
More Information
Set Membership: |
|

Tmem25
Genomic Map