Assay Details
| Gene Symbol: Tomm70a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645376.1 | XP_007643566.1 | 7 - 8 | 1053 | 70 | |
| GenBank mRNA | JI895616.1 | - | 7 - 8 | 1050 | 70 | |
More Information
Set Membership: |
|

Tomm70a
Genomic Map