Assay Details
| Gene Symbol: Psmb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498674.2 | XP_003498722.1 | 3 - 4 | 538 | 63 | |
| GenBank mRNA | JI874243.1 | - | 3 - 4 | 515 | 63 | |
| JP043174.1 | - | 3 - 4 | 516 | 63 | ||
More Information
Set Membership: |
|

Psmb4
Genomic Map