Assay Details
| Gene Symbol: Abhd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502730.2 | XP_003502778.1 | 6 - 7 | 802 | 65 | |
| XM_007643347.1 | XP_007641537.1 | 8 - 9 | 979 | 65 | ||
| XM_007643348.1 | XP_007641538.1 | 8 - 9 | 966 | 65 | ||
| XM_007643349.1 | XP_007641539.1 | 8 - 9 | 918 | 65 | ||
| XM_007643350.1 | XP_007641540.1 | 7 - 8 | 867 | 65 | ||
| XM_007643351.1 | XP_007641541.1 | 6 - 7 | 853 | 65 | ||
| GenBank mRNA | JI872017.1 | - | 6 - 7 | 848 | 65 | |
More Information
Set Membership: |
|

Abhd6
Genomic Map