Assay Details
| Gene Symbol: Pnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509215.2 | XP_003509263.1 | 1 - 2 | 179 | 59 | |
| GenBank mRNA | JP056140.1 | - | 1 - 2 | 88 | 59 | |
More Information
Set Membership: |
|

Pnp
Genomic Map