Assay Details
| Gene Symbol: Krt15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511329.1 | XP_003511377.1 | 1 - 2 | 494 | 93 | |
| XM_007651860.1 | XP_007650050.1 | 1 - 2 | 494 | 93 | ||
| XM_007651861.1 | XP_007650051.1 | 2 - 3 | 455 | 93 | ||

Krt15
Genomic Map