Assay Details
| Gene Symbol: Krt15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511329.1 | XP_003511377.1 | 6 - 7 | 1242 | 99 | |
| XM_007651860.1 | XP_007650050.1 | 6 - 7 | 1242 | 87 | ||
More Information
Set Membership: |
|

Krt15
Genomic Map